missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Multiparameter Chemical Test Kit
Click to view available options
For Use With (Application):
Cooling and Boiler Monitoring
Environmental Monitoring
Marine/Aquaculture Monitoring
Water Quality Monitoring
Unit Size:
Each
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, a case and pre-measured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
| For Use With (Application) | Marine/Aquaculture Monitoring |
| For Use With (Equipment) | Water samples |
| Includes | All necessary reagents and accessories for each test, pHep™ electronic pH tester, hard carrying case, and instructions |
| No. of Tests | 6 parameters x 100 tests each |
| Quantity | 1 Kit |
| Range | Various |
| Sufficient For | Testing of: alkalinity, carbon dioxide, hardness, dissolved oxygen, pH, and salinity |
| Test Method | Titration, colorimetry, and direct measurement |
| Type | Chemical Test Kit |
Safety and Handling
Product Identifier
- MARINE SCIENCE TEST KIT
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- H412-Harmful to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- EUH032-Contact with acids liberates very toxic gas.
- EUH071-Corrosive to the respiratory tract.
- EUH210-Safety data sheet available on request.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use powder to extinguish.
- P390-Absorb spillage to prevent material damage.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contains : Sodium hydroxide, Potassium iodide, Sodium azide, Propionicacid, Ethanol, Hydrochloric acid, Mercur
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction