missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Multiparameter Chemical Test Kit

Product Code. 11672360
Click to view available options
For Use With (Application):
Cooling and Boiler Monitoring
Environmental Monitoring
Marine/Aquaculture Monitoring
Water Quality Monitoring
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11672360

Brand: Hanna Instruments™ HI3817

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Multiparameter Chemical Test Kit features pre-made, pre-measured titration reagents for the determination of multiple parameters commonly measured for environmental/facility monitoring.

These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.

  • Complete setup: All required materials are included, such as beakers, syringes, a case and pre-measured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted

Specifications

For Use With (Application) Water Quality Monitoring
For Use With (Equipment) Water Samples
Includes All necessary reagents and accessories for each test, pHep™ electronic pH tester, hard carrying case, and instructions
No. of Tests 5 parameters x 100 tests, 1 parameter (iron) x 50 tests
Quantity 1 Kit
Range Various
Sufficient For Testing of: alkalinity, chloride, hardness, iron, pH, and sulfite
Test Method Titration, colorimetry, and direct measurement
Type Chemical Test Kit
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • water test kit
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Acute toxicity Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Germ cell mutagenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H290-May be corrosive to metals.
  • H373-May cause damage to organs.
  • H372-Causes damage to organs.,Causes damage to organs.
  • H302-Harmful if swallowed.,Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.,Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.,Suspected of causing genetic defects.
  • H319-Causes serious eye irritation.,Causes serious eye irritation.
  • H311-Toxic in contact with skin.,Toxic in contact with skin.
  • H318-Causes serious eye damage.,Causes serious eye damage.
  • H361f-Suspected of damaging fertility.,Suspected of damaging fertility.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.,Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H332-Harmful if inhaled.,Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.,Harmful to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.,May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.,Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.,Toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P391-Collect spillage.,Collect spillage.
  • P243-Take precautionary measures against static discharge.,Take precautionary measures against static discharge.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.,IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.,IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.,Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use powder to extinguish.,In case of fire: Use powder to extinguish.
  • P310-Immediately call a POISON CENTER/doctor/.,Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.,IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P235-Store in a well-ventilated place. Keep cool.,Store in a well-ventilated place. Keep cool.
  • P390-Absorb spillage to prevent material damage.,Absorb spillage to prevent material damage.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.,IF exposed or concerned: Get medical advice/attention.
  • P404-Store in a closed container.,Store in a closed container.
  • P337+P313-If eye irritation persists: Get medical advice/attention.,If eye irritation persists: Get medical advice/attention.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, phenolphtalein, potassium disulphate, sulfanilic acid, cadmium, 2,5-dihydroxybenzoic acid,
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.