Learn More
DWK Life Sciences DURAN™ Multi-purpose Cylinder, with round base, without graduation
Cylinders without graduation can be used to mix components.
Brand:  DWK Life Sciences 213985304
Additional Details : Weight : 3.30000kg
 View more versions of this product
Product Code. 15241729
  • 224.00€ / Pack of 10
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
  • Rough ground rim
  • With round base
Specifications
Specifications
Borosilicate Glass (3.3)
Diluting solutions; Mixing several components
Yes
50 mm
450 mL
10/Pk.