missing translation for 'onlineSavingsMsg'
Learn More

Mupirocin, 92%, Thermo Scientific™

406.00€ - 625.65€

Chemical Identifiers

CAS 12650-69-0
Molecular Formula C26H44O9
Molecular Weight (g/mol) 500.629
MDL Number MFCD01711620
InChI Key MINDHVHHQZYEEK-HBBNESRFSA-N
PubChem CID 446596
ChEBI CHEBI:7025
IUPAC Name 9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
SMILES CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15437669
SDS View Documents
Thermo Scientific Alfa Aesar
J66347.MF
50mg
406.00€
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15427669
SDS View Documents
Thermo Scientific Alfa Aesar
J66347.MC
100mg
ONLINE EXCLUSIVE PRICE
695.00€
625.65€ / 100mg
Save 69.35€
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Mupirocin is involved in inhibition of protein synthesis was reversed by isoleucine and Pseudomonic Acid was shown to inhibit IleRS (isoleucyl-tRNA synthetase) in vitro. It also demonstrates weak inhibitory action on FARSL (phenylalanyl-tRNA synthetase). Efficacy against staphylococci and streptococci including multiply resistant Staphylcoccus aureus and Staphylococcus epidermidis is reported with Pseudomonic Acid.

Solubility
Soluble in DMSO or methanol. Sparingly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Typical working concentrations are in the range of 0.25µg/mL-0.39µg/mL.
Specifications

Chemical Identifiers

12650-69-0
500.629
MINDHVHHQZYEEK-HBBNESRFSA-N
CHEBI:7025
CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O
C26H44O9
MFCD01711620
446596
9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid

Specifications

12650-69-0
>110°C (230°F)
MFCD01711620
14,6302
MINDHVHHQZYEEK-HBBNESRFSA-N
9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoic acid
446596
500.62
Mupirocin
White
C26H44O9
672°C
Soluble in DMSO or methanol; Sparingly soluble in water
CC(C1C(O1)CC2COC(C(C2O)O)CC(=CC(=O)OCCCCCCCCC(=O)O)C)C(C)O
500.629
CHEBI:7025
92%
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : RA6907000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.