Learn More
Cytiva Whatman™ Funnel Assembly for use with MV050 Vacuum Filtration Manifolds
Click to view available options
:
MV050 Funnel Assembly; Capacity: 500 mL
Description
- Product Type: Funnel Assembly
- Material: Stainless Steel
- Volume: 500 mL
- For Use with (Equipment): MV050 Vacuum Filtration Manifolds
- For Use with (Applications): Microbiology, Biochemistry, and Hydrobiology, Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
Specifications
Specifications
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | MV050 Manifold |
| Material | Stainless Steel |
| Sterility | Non-sterile |
| For Use With (Application) | For Use with (Applications): Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries) |
| Model | MV050/0/01 |
| Quantity | 1 Ea. |
| Series | MV050 |
| Type | Funnel Assembly |
| Show More |