Learn More
Metro™ MW Series Utility Cart with Wire Shelves
Made of Super Erecta™ shelves with plastic split sleeves, two one-piece handles and casters with donut bumpers, Metro MW Series Utility Cart with Wire Shelves make a smart addition to any workplace.
Brand: Metro™ MW711
Additional Details : Weight : 0.80000kg
Description
Cart
- Load capacity: 375 lb. (170.1kg)
- Fully interchangeable Super Erecta shelves
- Center shelf can be positioned at any height in 1 in. (2.5cm) increments
- One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability
- 18 in. (46cm) carts have 4 in. dia. (10cm) casters; 21 in.(53cm) and 24 in. (61cm) wide carts have 5 in. dia. (13cm) casters
- Three tiers
- 375 lbs (170kg) weight capacity per cart
- Chrome-plated or stainless-steel shelves available with trademarked truss design for added strength
- Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place
Order Info
Shipped knocked down. Assembles/disassembles quickly and easily without tools.
Specifications
Utility Cart | |
3-tier Model | |
Transporting materials that require an expanse of flat, solid surface | |
39 in. | |
Super Erecta™ 3 Chrome plated wire shelves with Plastic split sleeves, 2 one-piece Chrome plated Handle, 4 in. dia. caster, 3.5 in. donut bumpers | |
91 cm | |
170 kg | |
Chrome | |
61 cm |
NSF | |
36 x 24 x 39 in. (91 x 61 x 99 cm) | |
Chrome | |
99 cm | |
36 in. | |
375 lb. | |
3 | |
24 in. |