Learn More
BrandTech™ VACUUBRAND™ MZ 2C NT Synchro Dry Chemistry Multitasking Vacuum System
GREENER_CHOICE
Product Code. 10414574
Click to view available options
Plug Type:
CEE European
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10414574
Brand: BrandTech™ VACUUBRAND™ 20732800
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Provides powerful oil-free vacuum for two independent applications, such as rotary evaporators, gel dryers or other common lab tasks. VACUUBRAND™ MZ2C NT Synchro Dry Chemistry Multitasking Vacuum System builds on the popular MZ2C NT diaphragm vacuum pump.
Compact and whisper-quiet, it builds on the popular MZ 2C NT diaphragm vacuum pump.
  • Ultimate vacuum: 5 Torr (7 mbar)
  • Free air capacity: 38 lpm (1.4 cfm)
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 5.3 torr (without Gas Ballast), 9 torr (with Gas Ballast)
Horsepower 0.24 hp
Includes Manual
Motor Speed 150 to 1800 rpm
Certifications/Compliance ATEX Conformity II 3G IIC T3 X, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010, NRTL Certification, TUV
Electrical Requirements 230 V
Material PTFE, Fluoroplastic (Case)
Model 732800 Vacuubrand MZ2C NT+AK Synchro+EK
Noise Level 45 dbA
Plug Type CEE European
Temperature (English) Max. 104°F
Flow Rate 1.2/1.4 CFM
Frequency 50/60 Hz
Overall Height (Metric) 401.32 mm
Overall Length (Metric) 325.12 mm
Overall Width (Metric) 248.92 mm
Pump Type Diaphragm Pump
Enclosure Rating IP40
No. of Pump Heads 2
Resistance Properties Chemical-Resistant
Temperature (English) Min 50°F
Applications Rotary Evaporation
Oil Type Oil-Free
No. of Stages 2
Maximum Pressure 16 psi
Voltage 230 V