Learn More
Macherey-Nagel™ N 12 Septa Replacement Screw Caps
Brand:  Macherey-Nagel™ 702292
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12794658
  • 24.51€ / Pack of 100
Estimated Shipment: 03-08-2021
Add to basket
Specifications
Red, White
Septa N 12
1.3 mm
Silicone, PTFE
100 Pack
Micro Reaction Vials N 13