missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl)methyl]-N-methylamine, ≥95%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 1-(2,2-dimethyl-3H-1-benzofuran-7-yl)-N-methylmethanamine |
Formula Weight | 191.27 |
Percent Purity | 95% |
Chemical Name or Material | N-[(2, 2-dimethyl-2, 3-dihydro-1-benzofuran-7-yl)methyl]-N-methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'