missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromobutyl)phthalimide, 98%, Thermo Scientific Chemicals

Catalog Number p-3735705
55.20 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5394-18-3
C12H12BrNO2
282.14
MFCD00005905
UXFWTIGUWHJKDD-UHFFFAOYSA-N
n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide
93575
2-(4-bromobutyl)isoindole-1,3-dione
BrCCCCN1C(=O)C2=CC=CC=C2C1=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5394-18-3
C12H12BrNO2
282.14
MFCD00005905
UXFWTIGUWHJKDD-UHFFFAOYSA-N
n-4-bromobutyl phthalimide, 2-4-bromobutyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-bromobutyl, 2-4-bromobutyl-1h-isoindole-1,3 2h-dione, n-4-bromobutyl-phthalimide, 2-4-bromobutyl isoindole-1,3-dione, 2-4-bromobutyl-2,3-dihydro-1h-isoindole-1,3-dione, n-4-bromobutyl, n-bromobutylphthalimide
93575
2-(4-bromobutyl)isoindole-1,3-dione
BrCCCCN1C(=O)C2=CC=CC=C2C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 76.0°C to 80.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 21, 463
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 282.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(4-Bromobutyl)phthalimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'