missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Cyanophenyl)guanidine, 97%, Thermo Scientific™

Catalog Number 15430968
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
5637-42-3
C8H8N4
160.18
MFCD16251323
BTDGLZSKNFJBER-UHFFFAOYSA-N
1-4-cyanophenyl guanidine, n-4-cyanophenyl guanidine, 2-4-cyanophenyl guanidine, 4-cyanophenyl guanidine, 4-cyanophenylguanidine
17747914
2-(4-cyanophenyl)guanidine
C1=CC(=CC=C1C#N)N=C(N)N
This item is not returnable. View return policy
5637-42-3
C8H8N4
160.18
MFCD16251323
BTDGLZSKNFJBER-UHFFFAOYSA-N
1-4-cyanophenyl guanidine, n-4-cyanophenyl guanidine, 2-4-cyanophenyl guanidine, 4-cyanophenyl guanidine, 4-cyanophenylguanidine
17747914
2-(4-cyanophenyl)guanidine
C1=CC(=CC=C1C#N)N=C(N)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material N-(4-Cyanophenyl)guanidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'