missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Cholecystokinin, CCK (26-31), non-sulfated, Thermo Scientific™

Catalog Number 15400687
Click to view available options
Quantity:
0.5 mg
1 mg
This item is not returnable. View return policy
MFCD00133562
N-Acetyl-Asp-Tyr-Met-Gly-Trp-Met-NH2
This item is not returnable. View return policy
MFCD00133562
N-Acetyl-Asp-Tyr-Met-Gly-Trp-Met-NH2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 0.5 mg
Formula Weight 842.98
Physical Form Solid
Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-31), non-sulfated
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'