missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylethanolamine (~90%), TRC
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
Specifications
Specifications
| Chemical Name or Material | N-Acetylethanolamine |
| CAS | 142-26-7 |
| Molecular Formula | C4 H9 N O2 |
| Quantity | 1 g |
| Synonym | N-(2-Hydroxyethyl)acetamide, 2-Acetamidoethanol, 2-Acetylaminoethanol, 2-N-Acetylaminoethanol, Acetamide MEA, Acetylcolamine, Incromectant AMEA 100, Incromectant AMEA 70, Mackamide AME 100, N-(β-Hydroxyethyl)acetamide, N-Acetyl-2-aminoethanol, N-Ethanolacetamide, NSC 5999, Schercomid AME 70, β-Hydroxyethylacetamide |
| InChI Formula | InChI=1S/C4H9NO2/c1-4(7)5-2-3-6/h6H,2-3H2,1H3,(H,5,7) |
| SMILES | CC(=O)NCCO |
| IUPAC Name | N-(2-hydroxyethyl)acetamide |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction