Learn More
Alfa Aesar™ N-Boc-2-bromobenzylamine, 97%
Brand: Alfa Aesar™ H55190.03
Additional Details : CAS : 162356-90-3 Weight : 0.00100kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
162356-90-3 | |
286.169 | |
DFNZFCPEUDSNEO-UHFFFAOYSA-N | |
7018300 | |
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br |
C12H16BrNO2 | |
MFCD05664385 | |
tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804 | |
tert-butyl N-[(2-bromophenyl)methyl]carbamate |
Specifications
N-Boc-2-bromobenzylamine | |
White to Brown | |
1g | |
C12H16BrNO2 | |
UN3077 | |
DFNZFCPEUDSNEO-UHFFFAOYSA-N | |
tert-butyl N-[(2-bromophenyl)methyl]carbamate | |
7018300 | |
Solid |
>110°C (230°F) | |
50°C to 54°C | |
162356-90-3 | |
MFCD05664385 | |
tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804 | |
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br | |
286.169 | |
286.17 | |
97% |