missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-cyclohexyl-L-glycine, 97%, Thermo Scientific™

Catalog Number p-7054340
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
109183-71-3
C13H23NO4
257.33
MFCD00133638
QSUXZIPXYDQFCX-JTQLQIEISA-N
boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid
7004938
(2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O
This item is not returnable. View return policy
109183-71-3
C13H23NO4
257.33
MFCD00133638
QSUXZIPXYDQFCX-JTQLQIEISA-N
boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid
7004938
(2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-2-cyclohexyl-L-glycine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Beilstein 5553687
Solubility Information Slightly soluble in water.
Formula Weight 257.33
Percent Purity 97%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Boc-2-cyclohexyl-L-glycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'