missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-bromo-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog Number p-7034343
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
82278-73-7
C14H18BrNO4
344.205
MFCD01317710
FBUDYESOPLBQIR-NSHDSACASA-N
s-n-boc-3-bromophenylalanine, boc-l-3-bromophenylalanine, boc-3-bromo-l-phenylalanine, boc-phe 3-br-oh, boc-l-3-br-phe-oh, boc-l-3-bromophe, n-boc-3-bromo-l-phenylalanine, s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-3-bromophenyl propanoic acid
2734476
(2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O
This item is not returnable. View return policy
82278-73-7
C14H18BrNO4
344.205
MFCD01317710
FBUDYESOPLBQIR-NSHDSACASA-N
s-n-boc-3-bromophenylalanine, boc-l-3-bromophenylalanine, boc-3-bromo-l-phenylalanine, boc-phe 3-br-oh, boc-l-3-br-phe-oh, boc-l-3-bromophe, n-boc-3-bromo-l-phenylalanine, s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-3-bromophenyl propanoic acid
2734476
(2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O

N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 344.2
Percent Purity 95%
Chemical Name or Material N-Boc-3-bromo-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'