Learn More
Alfa Aesar™ N-Boc-3-cyano-L-phenylalanine, 95%
Brand: Alfa Aesar™ H62517.MD
Additional Details : CAS : 131980-30-8 Weight : 0.00025kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 250mg |
---|
Chemical Identifiers
131980-30-8 | |
290.319 | |
FDQDHMZKOPOWFE-LBPRGKRZSA-N | |
2734498 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O |
C15H18N2O4 | |
MFCD00797560 | |
boc-phe 3-cn-oh, boc-l-3-cyanophenylalanine, boc-3-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-l-3-cn-phe-oh, 2s-2-tert-butoxy carbonylamino-3-3-cyanophenyl propanoic acid, boc-l-3-cyanophe, boc-l-3-cyano phenylalanine | |
(2S)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
N-Boc-3-cyano-L-phenylalanine | |
C15H18N2O4 | |
UN3439 | |
FDQDHMZKOPOWFE-LBPRGKRZSA-N | |
(2S)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2734498 | |
Solid | |
250mg |
131980-30-8 | |
MFCD00797560 | |
boc-phe 3-cn-oh, boc-l-3-cyanophenylalanine, boc-3-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid, boc-l-3-cn-phe-oh, 2s-2-tert-butoxy carbonylamino-3-3-cyanophenyl propanoic acid, boc-l-3-cyanophe, boc-l-3-cyano phenylalanine | |
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O | |
290.319 | |
290.31 | |
95% |