missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-glutamic acid 5-benzyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-7041545
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
35793-73-8
C17H23NO6
337.372
MFCD00038258
AJDUMMXHVCMISJ-CYBMUJFWSA-N
boc-d-glu obzl-oh, boc-d-glutamic acid 5-benzyl ester, boc-d-glutamic acid 5-benzylester, n-boc-d-glutamic acid 5-benzyl ester, 2r-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, ambotzbaa1354, boc-d-glutamic acid gamma-benzyl ester, pubchem12172, pubchem14934, boc-d-glu obn-oh
7010513
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O
This item is not returnable. View return policy
35793-73-8
C17H23NO6
337.372
MFCD00038258
AJDUMMXHVCMISJ-CYBMUJFWSA-N
boc-d-glu obzl-oh, boc-d-glutamic acid 5-benzyl ester, boc-d-glutamic acid 5-benzylester, n-boc-d-glutamic acid 5-benzyl ester, 2r-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, ambotzbaa1354, boc-d-glutamic acid gamma-benzyl ester, pubchem12172, pubchem14934, boc-d-glu obn-oh
7010513
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 337.37
Percent Purity 98%
Chemical Name or Material N-Boc-D-glutamic acid 5-benzyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'