missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine tert-butyl ester, 95%, Thermo Scientific Chemicals

Catalog Number 15408908
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
111652-20-1
C11H21NO4
231.29
MFCD00191866
ZDKFMHKWZATBMR-UHFFFAOYSA-N
boc-gly-otbu, tert-butyl 2-tert-butoxycarbonyl amino acetate, boc-glycine tert-butyl ester, n-boc-glycine tert-butyl ester, n-tert-butoxycarbonyl glycine tert-butyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, 1,1-dimethylethyl ester, tert-butyl 2-2-methylpropan-2-yl oxycarbonylamino acetate, 2-azabutanedioic acid di-tert-butyl ester
4097102
CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C
This item is not returnable. View return policy
111652-20-1
C11H21NO4
231.29
MFCD00191866
ZDKFMHKWZATBMR-UHFFFAOYSA-N
boc-gly-otbu, tert-butyl 2-tert-butoxycarbonyl amino acetate, boc-glycine tert-butyl ester, n-boc-glycine tert-butyl ester, n-tert-butoxycarbonyl glycine tert-butyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, 1,1-dimethylethyl ester, tert-butyl 2-2-methylpropan-2-yl oxycarbonylamino acetate, 2-azabutanedioic acid di-tert-butyl ester
4097102
CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 66°C to 68°C
Linear Formula (CH3)3COCONHCH2CO2C(CH3)3
Quantity 25 g
Beilstein 5002580
IUPAC Name tert-butyl 2-{[(tert-butoxy)carbonyl]amino}acetate
Formula Weight 231.29
Percent Purity 95%
Chemical Name or Material N-Boc-glycine tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'