missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-prolinol, 98+%

Brand:  Alfa Aesar™ L09885.06

Additional Details : CAS : 69610-40-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408919

  • 71.03€ / 5g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

69610-40-8
201.266
BFFLLBPMZCIGRM-QMMMGPOBSA-N
643448
CC(C)(C)OC(=O)N1CCCC1CO
C10H19NO3
MFCD00066232
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Specifications

N-Boc-L-prolinol
59°C to 62°C
69610-40-8
MFCD00066232
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
CC(C)(C)OC(=O)N1CCCC1CO
201.266
201.27
−54° (c=5 in Methanol)
5g
C10H19NO3
3542667
BFFLLBPMZCIGRM-QMMMGPOBSA-N
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
643448
≥98%
Videos
SDS Documents