missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cbz-L-aspartic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N-Cbz-L-aspartic Acid |
| CAS | 1152-61-0 |
| Molecular Formula | C12H13NO6 |
| Quantity | 10 g |
| Synonym | N-[(Phenylmethoxy)carbonyl]-L-aspartic Acid, N-Carboxy-L-aspartic Acid N-Benzyl Ester, Benzyloxycarbonyl-L-aspartic Acid, Carbobenzoxy-L-aspartic Acid, Cbz-Asp-OH, N-(Benzyloxycarbonyl)-L-aspartic Acid, N-Benzyloxylcarbonyl-L-aspartic Acid, N-Carbobenzoxy-L-aspartic Acid, N-Carbobenzyloxy-L-aspartic Acid, NSC 88479, NSC 9972 |
| InChI Formula | InChI=1S/C12H13NO6/c14-10(15)6-9(11(16)17)13-12(18)19-7-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,18)(H,14,15)(H,16,17)/t9-/m0/s1 |
| SMILES | OC(=O)C[C@H](NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-2-(phenylmethoxycarbonylamino)butanedioic acid |
| Molecular Weight (g/mol) | 267.23 |
| Formula Weight | 267.0743 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction