missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ n-Dodecyltrichlorosilane, 96%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
n-Dodecyltrichlorosilane is employed in the medicinal field, electronic industry and used as polymer material. It is a coupling and reducing agent. It is also used as a lubricant.
Solubility
Soluble in parts of organic solvents.
Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.
n-Dodecyltrichlorosilane is employed in the medicinal field, electronic industry and used as polymer material. It is a coupling and reducing agent. It is also used as a lubricant.
Solubility
Soluble in parts of organic solvents.
Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.
Specifications
Boiling Point | 155°C (10mmHg) |
Quantity | 25 g |
Linear Formula | CH3(CH2)11SiCl3 |
UN Number | UN1771 |
Beilstein | 1758004 |
Flash Point | 165°C (329°F) |
Solubility Information | Soluble in parts of organic solvents. |
Formula Weight | 303.78 |
Percent Purity | 96% |
Refractive Index | 1.458 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'