missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylisopropylamine, TRC

Product Code. 30377149
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377149 5 g 5g
30376744 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30377149 Supplier Toronto Research Chemicals Supplier No. TRCE9219505G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethylisopropylamine
CAS 19961-27-4
Molecular Formula C5H13N
Quantity 5 g
Synonym N-Ethyl-2-propanamine, 1-Methyldiethylamine, (Ethyl)(1-methylethyl)amine, 2-(Ethylamino)propane, Ethylisopropylamine, Isopropylethylamine, N-Ethyl-2-propanamine, N-Ethyl-N-isopropylamine, NSC 165659
InChI Formula InChI=1S/C5H13N/c1-4-6-5(2)3/h5-6H,4H2,1-3H3
SMILES CCNC(C)C
IUPAC Name N-ethylpropan-2-amine
Molecular Weight (g/mol) 87.16
Formula Weight 87.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.