missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid 4-methyl ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7055245
44.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
145038-53-5
C20H19NO6
369.373
MFCD08458573
HSOKCYGOTGVDHL-KRWDZBQOSA-N
fmoc-asp ome-oh, fmoc-l-aspartic acid 4-methyl ester, fmoc-aspartic acid 4-methyl ester, fmoc-l-asp ome-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid 4-methyl ester, n-, a-fmoc-l-aspartic acid, a-methyl ester
45382185
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methoxy-4-oxobutanoic acid
COC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
145038-53-5
C20H19NO6
369.373
MFCD08458573
HSOKCYGOTGVDHL-KRWDZBQOSA-N
fmoc-asp ome-oh, fmoc-l-aspartic acid 4-methyl ester, fmoc-aspartic acid 4-methyl ester, fmoc-l-asp ome-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid 4-methyl ester, n-, a-fmoc-l-aspartic acid, a-methyl ester
45382185
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methoxy-4-oxobutanoic acid
COC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 369.37
Percent Purity 97%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Fmoc-L-aspartic acid 4-methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'