missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-D-tyrosine, 95%, Thermo Scientific Chemicals

Catalog Number p-7041600
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
118488-18-9
C28H29NO5
459.54
MFCD00065684
JAUKCFULLJFBFN-KSYWNVGFNA-N
fmoc-d-tyr tbu-oh, fmoc-o-tert-butyl-d-tyrosine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, nalpha-9h-fluoren-9-ylmethoxy carbonyl-o-tert-butyl-d-tyrosine, 2r-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-tyrosine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, ambotzfaa1341, pubchem10054, fmoc-d-tyr but-oh, fmoc-o-tert.butyl-d-tyrosine
12968135
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
This item is not returnable. View return policy
118488-18-9
C28H29NO5
459.54
MFCD00065684
JAUKCFULLJFBFN-KSYWNVGFNA-N
fmoc-d-tyr tbu-oh, fmoc-o-tert-butyl-d-tyrosine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, nalpha-9h-fluoren-9-ylmethoxy carbonyl-o-tert-butyl-d-tyrosine, 2r-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-tyrosine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, ambotzfaa1341, pubchem10054, fmoc-d-tyr but-oh, fmoc-o-tert.butyl-d-tyrosine
12968135
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

N-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-O-tert-butyl-D-tyrosine is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

Solubility
Soluble in 1 mmole in 2 ml dimethylformamide (Clearly soluble). Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 6691868
Solubility Information Soluble in 1 mmole in 2 ml dimethylformamide (Clearly soluble). Slightly soluble in water.
Formula Weight 459.53
Percent Purity 95%
Chemical Name or Material N-Fmoc-O-tert-butyl-D-tyrosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'