missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Formylpiperidine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -31°C |
Color | Colorless to Yellow |
Density | 1.0190g/mL |
Boiling Point | 222°C |
Flash Point | 91°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- N-Formylpiperidine
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'