missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexane, dried (max. 100 ppm water), Honeywell Burdick & Jackson™
Click to view available options
185 L
Specifications
Melting Point | -95°C |
CAS | 110-54-3 |
Color | Colorless |
CAS Max % | 1 |
Molecular Formula | C6H14 |
Linear Formula | CH3(CH2)4CH3 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
SMILES | CCCCCC |
IUPAC Name | hexane |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'