missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxy Vildagliptin, TRC

Product Code. 30451778
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 30451778

Brand: Toronto Research Chemicals TRCV305050100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Hydroxy Vildagliptin
Analyte or Component Names N-Hydroxy Vildagliptin
Shipping Condition Room Temperature
Molecular Formula C17 H25 N3 O3
InChI Formula InChI=1S/C17H25N3O3/c18-9-14-2-1-3-19(14)15(21)10-20(23)16-5-12-4-13(6-16)8-17(22,7-12)11-16/h12-14,22-23H,1-8,10-11H2/t12-,13+,14-,16+,17-/m0/s1
Solution Type Neat
SMILES ON(CC(=O)N1CCC[C@H]1C#N)[C@@]23C[C@@H]4C[C@@H](C[C@@](O)(C4)C2)C3
IUPAC Name (2S)-1-[2-[hydroxy-[(5S,7R)-3-hydroxy-1-adamantyl]amino]acetyl]pyrrolidine-2-carbonitrile
Industry Type Pharmaceutical
Molecular Weight (g/mol) 319.399
Quantity 100 mg
Formula Weight 319.19
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.