missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[1-(6-methylpyrazin-2-yl)piperid-3-yl]methylamine, 97%, Thermo Scientific™

Product Code. p-3586868
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10205384

Brand: Thermo Scientific Maybridge CC59046CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 941716-84-3
Molecular Formula C12H20N4
Molecular Weight (g/mol) 220.32
MDL Number MFCD09879920
InChI Key CBRQUNBWUQBINE-UHFFFAOYSA-N
Synonym n-methyl-1-6-methylpyrazin-2-yl piperid-3-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-3-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 3-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-3-yl methanamine
PubChem CID 24229603
IUPAC Name N-methyl-1-[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanamine
SMILES CC1=CN=CC(=N1)N2CCCC(C2)CNC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 220.32
Percent Purity 97%
Chemical Name or Material N-Methyl-[1-(6-methylpyrazin-2-yl)piperid-3-yl]methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.