missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-propanamine, TRC

Product Code. 30468344
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468344 10 g 10g
30482850 25 g 25g
30483486 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468344 Supplier Toronto Research Chemicals Supplier No. TRCM32630510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-35-0
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
Synonym Methylpropylamine, N-Methyl-1-propanamine, N-Methyl-1-propylamine, N-Methyl-n-propylamine, N-Methylpropan-1-ylamine, N-Methylpropanamine, N-Methylpropylamine, NSC 165652, Propylmethylamine, n-Propylmethylamine
IUPAC Name N-methylpropan-1-amine
SMILES CCCNC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H11N/c1-3-4-5-2/h5H,3-4H2,1-2H3
Formula Weight 73.0891
Chemical Name or Material N-Methyl-1-propanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.