missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-propanamine, TRC

Product Code. 30483486
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
Product Code. Quantity unitSize
30483486 5 g 5g
30468344 10 g 10g
30482850 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 30483486

Brand: Toronto Research Chemicals TRCM3263055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-35-0
Formula Weight 73.0891
InChI Formula InChI=1S/C4H11N/c1-3-4-5-2/h5H,3-4H2,1-2H3
IUPAC Name N-methylpropan-1-amine
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
SMILES CCCNC
Synonym Methylpropylamine, N-Methyl-1-propanamine, N-Methyl-1-propylamine, N-Methyl-n-propylamine, N-Methylpropan-1-ylamine, N-Methylpropanamine, N-Methylpropylamine, NSC 165652, Propylmethylamine, n-Propylmethylamine
Chemical Name or Material N-Methyl-1-propanamine

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.