missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-phenoxybenzylamine, 97%, Thermo Scientific™

Catalog Number p-3175636
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
129535-78-0
C14H15NO
213.28
MFCD02089413
GDVLOOQWWBPGSV-UHFFFAOYSA-N
n-methyl-3-phenoxybenzylamine, n-methyl-1-3-phenoxyphenyl methanamine, methyl 3-phenoxyphenyl methyl amine, n-methyl-3-phenoxybenzenemethanamine, benzenemethanamine, n-methyl-3-phenoxy, acmc-209bgx, benzenemethanamine,n-methyl-3-phenoxy
10512764
N-methyl-1-(3-phenoxyphenyl)methanamine
CNCC1=CC(=CC=C1)OC2=CC=CC=C2
This item is not returnable. View return policy
129535-78-0
C14H15NO
213.28
MFCD02089413
GDVLOOQWWBPGSV-UHFFFAOYSA-N
n-methyl-3-phenoxybenzylamine, n-methyl-1-3-phenoxyphenyl methanamine, methyl 3-phenoxyphenyl methyl amine, n-methyl-3-phenoxybenzenemethanamine, benzenemethanamine, n-methyl-3-phenoxy, acmc-209bgx, benzenemethanamine,n-methyl-3-phenoxy
10512764
N-methyl-1-(3-phenoxyphenyl)methanamine
CNCC1=CC(=CC=C1)OC2=CC=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 213.28
Percent Purity 97%
Chemical Name or Material N-Methyl-3-phenoxybenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'