missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-(4-pyridylmethyl)amine, 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 122.17 |
Percent Purity | 97% |
Chemical Name or Material | N-Methyl-N-(4-pyridinylmethyl)amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'