missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals

Catalog Number p-7020992
Click to view available options
Quantity:
5 g
25 g
100 g
24589-78-4
C6H12F3NOSi
199.25
MFCD00000411
MSPCIZMDDUQPGJ-UHFFFAOYSA-N
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
32510
CHEBI:85064
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
CN(C(=O)C(F)(F)F)[Si](C)(C)C
24589-78-4
C6H12F3NOSi
199.25
MFCD00000411
MSPCIZMDDUQPGJ-UHFFFAOYSA-N
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
32510
CHEBI:85064
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
CN(C(=O)C(F)(F)F)[Si](C)(C)C

N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Density 1.076
Boiling Point 130°C to 132°C
Flash Point 25°C (77°F)
Refractive Index 1.38
Quantity 5 g
UN Number UN1993
Beilstein 1941550
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)-Acetamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'