missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Burdick & Jackson™ N,N-Diisopropylethylamine, Biotech Grade, 99.5%, Honeywell™ Burdick & Jackson™

Biotech Grade, 99.5%

Brand:  Honeywell Burdick & Jackson™ 496219-500ML

Additional Details : CAS : 7087-68-5 Poids : 0.37100kg Transport : UN number : 3384 Chem class : 6.1 Pack group : I

Packaging Glass Bottle
Quantity 500mL

 View more versions of this product

Product Code. 15655690

  • 306.44€ / 500mL


Expédition estimée
10-02-2021
Add to basket

Description

Description

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Specifications

Chemical Identifiers

7087-68-5
129.247
JGFZNNIVVJXRND-UHFFFAOYSA-N
81531
CCN(C(C)C)C(C)C
C8H19N
MFCD00008868
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
N-ethyl-N-propan-2-ylpropan-2-amine

Specifications

N,N-Diisopropylethylamine
12°C (53.6°F)
Glass Bottle
Undesignated
500mL
C8H19N
MFCD00008868
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
CCN(C(C)C)C(C)C
129.247
129.24g/mol
0.742g/mL at 25°C
Strong, amine like
128.33°C
-19.99°C
7087-68-5
[(CH3)2CH]2NC2H5
UN3384
JGFZNNIVVJXRND-UHFFFAOYSA-N
N-ethyl-N-propan-2-ylpropan-2-amine
81531
99.50%
Videos
Documents