Learn More
N,N-Diisopropylethylamine, Biotech Grade, 99.5%, Honeywell Burdick & Jackson™
Catalog Number p-7120609
Click to view available options
Quantity:
100 mL
500 mL
2 L
Packaging:
Glass Bottle
7087-68-5
C8H19N
129.247
MFCD00008868
JGFZNNIVVJXRND-UHFFFAOYSA-N
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
81531
N-ethyl-N-propan-2-ylpropan-2-amine
CCN(C(C)C)C(C)C
7087-68-5
C8H19N
129.247
MFCD00008868
JGFZNNIVVJXRND-UHFFFAOYSA-N
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
81531
N-ethyl-N-propan-2-ylpropan-2-amine
CCN(C(C)C)C(C)C
Biotech Grade, 99.5%
  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY
Melting Point -19.99°C
Density 0.742g/mL at 25°C
Boiling Point 128.33°C
Flash Point 12°C (53.6°F)
Odor Strong, amine like
Packaging Glass Bottle
Linear Formula [(CH3)2CH]2NC2H5
Quantity 100 mL
UN Number UN3384
Formula Weight 129.24g/mol
Percent Purity 99.50%
Chemical Name or Material N,N-Diisopropylethylamine