missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals

Catalog Number p-3757494
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1188-33-6
C7H17NO2
147.22
MFCD00009227
BWKAYBPLDRWMCJ-UHFFFAOYSA-N
n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
70913
1,1-diethoxy-N,N-dimethylmethanamine
CCOC(N(C)C)OCC
This item is not returnable. View return policy
1188-33-6
C7H17NO2
147.22
MFCD00009227
BWKAYBPLDRWMCJ-UHFFFAOYSA-N
n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
70913
1,1-diethoxy-N,N-dimethylmethanamine
CCOC(N(C)C)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Density 0.8590g/mL
Boiling Point 128°C to 130°C
Flash Point 28°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2NCH(OC2H5)2
Quantity 10 g
Fieser 01,281; 02,154; 05,253; 07,125; 10,157
Specific Gravity 0.859
Solubility Information Solubility in water: decomposes
Formula Weight 147.22
Percent Purity 95%
Chemical Name or Material N,N-Dimethylformamide diethyl acetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'