missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzohydroxamic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3779415
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
304-88-1
C13H11NO2
213.24
MFCD00002111
YLYIXDZITBMCIW-UHFFFAOYSA-N
n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid
67536
N-hydroxy-N-phenylbenzamide
ON(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
304-88-1
C13H11NO2
213.24
MFCD00002111
YLYIXDZITBMCIW-UHFFFAOYSA-N
n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid
67536
N-hydroxy-N-phenylbenzamide
ON(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 119°C to 123°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CON(OH)C6H5
Quantity 5 g
Formula Weight 213.24
Percent Purity 98%
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material N-Phenylbenzohydroxamic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'