missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-t-Butyl N-Deactyl Linezolid, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | (5S)-5-[[(1,1-Dimethylethyl)amino]methyl]-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone |
| CAS | 1215006-08-8 |
| Molecular Formula | C18 H26 F N3 O3 |
| Quantity | 5 mg |
| Synonym | (5S)-5-[[(1,1-Dimethylethyl)amino]methyl]-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone, (-)-(S)-5-[(tert-Butylamino)methyl]-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one |
| InChI Formula | InChI=1S/C18H26FN3O3/c1-18(2,3)20-11-14-12-22(17(23)25-14)13-4-5-16(15(19)10-13)21-6-8-24-9-7-21/h4-5,10,14,20H,6-9,11-12H2,1-3H3/t14-/m0/s1 |
| SMILES | CC(C)(C)NC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2 |
| IUPAC Name | (5S)-5-[(tert-butylamino)methyl]-3-(3-fluoro-4-morpholin-4-ylphenyl)-1,3-oxazolidin-2-one |
| Molecular Weight (g/mol) | 351.42 |
| Formula Weight | 351.1958 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction