Learn More
Aalborg™ N2 Mass Flowmeter
Product Code. 11892370
Click to view available options
Flow Rate Range:
0 to 2 LPM
0 to 20 LPM
0 to 30 LPM
0 to 50 LPM
0 to 500 mL/min.
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11892370
Brand: Aalborg™ GFM37AVAL6A0/3NC33SS
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Type Mass Flowmeter
Accuracy ±5%
Flow Rate Range 0 to 50 LPM
Power Supply Details 12 V to 200 mA max
Precision ±1.5 %
Scale Full scale
Temperature (Metric) 5°C to 50°C