Learn More
Invitrogen™ NA-Star™ Detection Microplates
Product Code. 12664255
Click to view available options
Quantity:
10 plates
Unit Size:
Pack of 10
Product Code. 12664255
Brand: Invitrogen™ 4374349
Solid white 96-well microplates, supplied with covers, optimized for use with the chemiluminescent assay reagents. Thermo Scientific NA-Star NA-Star Detection Microplates are ideal for use with other chemiluminescent assays that do not require a specific
Ideal for use with other chemiluminescent assays that do not require a specific surface chemistry or coating. They do provide a high binding surface that can be coated with specific capture reagents for use with immunoassay⁄capture assay applications, if desired.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Content And Storage Contains 10 plates with covers
Detection Method Colorimetric
Product Line NA-Star NovaBright
For Use With (Equipment) Luminometer (Microplate)
Plate Blocking None
Quantity 10 plates
Shipping Condition Room Temperature