missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naftidrofuryl Hydrogen Oxalate, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Naftidrofuryl Hydrogen Oxalate
Specifications
Specifications
| Chemical Name or Material | Naftidrofuryl Hydrogen Oxalate |
| CAS | 6-4-3200 |
| Analyte or Component Names | Naftidrofuryl Hydrogen Oxalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H33 N O3 . C2 H2 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Naftidrofuryl Hydrogen Oxalate, Citoxid, Di-Actane, Dubimax, Dusodril, EU 1806, Iridus, LS 121, NSC 225233, Nafronyl acid oxalate, Nafronyl oxalate, Naftidrofuryl oxalate, Naphtidrofuryl oxalate, Praxilene, 2-Furanpropanoic acid, tetrahydro-alpha-(1-naphthalenylmethyl)-, 2-(diethylamino)ethyl ester, ethanedioate (1:1), 2-Furanpropionic acid, tetrahydro-alpha-(1-naphthylmethyl)-, 2-(diethylamino)ethyl ester oxalate (1:1) (8CI), 2-Furanpropionic acid, tetrahydro-α-(1-naphthylmethyl)-, 2-(diethylamino)ethyl ester oxalate (1:1) (8CI) |
| InChI Formula | InChI=1S/C24H33NO3.C2H2O4/c1-3-25(4-2)14-16-28-24(26)21(18-22-12-8-15-27-22)17-20-11-7-10-19-9-5-6-13-23(19)20;3-1(4)2(5)6/h5-7,9-11,13,21-22H,3-4,8,12,14-18H2,1-2H3;(H,3,4)(H,5,6) |
| SMILES | CCN(CC)CCOC(=O)C(CC1CCCO1)Cc2cccc3ccccc23.OC(=O)C(=O)O |
| IUPAC Name | 2-(diethylamino)ethyl 2-(naphthalen-1-ylmethyl)-3-(oxolan-2-yl)propanoate;oxalic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction