Learn More
Rogo Sampaic™ Nageotte Counting Cells
Product Code. 11770835
Click to view available options
Unit Size:
Each
Product Code. 11770835
Brand: Rogo Sampaic™ ZMB054
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For determining the number of particles per volume unit of a liquid. Rogo Sampaic™ Nageotte Counting Cells have a simple or double grid with a depth of 0.5mm.
  • For spinal and urinary counts
  • Provides for accurate manual measurements
Specifications
Product Type Nageotte Cell
Grid Type Simple
Quantity 1Each
Sample Type Spinal, Urinary
Depth (Metric) 0.5mm