missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Nonsterile, in Tray Pack

Catalog Number 10612634
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Store and ship small-volume, light-sensitive samples using these diagnostic bottles, which are tray packed for easy handling. These bottles can be used in test kits and many small-volume applications.

Store and ship small-volume, light-sensitive samples using Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. Used in test kits and many small volume applications, the opaque amber color of these high-density polyethylene bottles make them ideal for packaging light-sensitive reagents. The durable, uniform walls are resistant to splitting or puncturing. Bottles and closures are packed in rigid shrink-wrapped trays, allowing for easy handling.

Features:

  • Nonsterile
  • Opaque amber high-density polyethylene with opaque amber polypropylene closure provides excellent chemical resistance
  • Leakproof†

Ordering Information:

Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.

Warranty: 90 day

TRUSTED_SUSTAINABILITY
Capacity 0.125 oz., 4 mL
Closure Material Polypropylene
Closure Size 13-415 mm
Closure Type Screw Cap
Diameter (Metric) 16 mm
Height (English) 1.61 in.
Height (Metric) 41 mm
Width (English) 0.62 in.
Product Type Diagnostic Bottle
Capacity (Metric) 4 mL
Capacity (English) 0.125 oz.
Color Opaque Amber
Diameter (Metric) Inner Neck 8 mm
Material HDPE
No. per Case 1328
No. per Pack 332
Sterility Non-sterile
Neck Finish 13-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Show More Show Less

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'