Learn More
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Nonsterile, in Tray Pack
Description
Store and ship small-volume, light-sensitive samples using Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. Used in test kits and many small volume applications, the opaque amber color of these high-density polyethylene bottles make them ideal for packaging light-sensitive reagents. The durable, uniform walls are resistant to splitting or puncturing. Bottles and closures are packed in rigid shrink-wrapped trays, allowing for easy handling.
- Nonsterile
- Opaque amber high-density polyethylene with opaque amber polypropylene closure provides excellent chemical resistance
- Leakproof†
Ordering Information:
Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.
Warranty: 90 day
Specifications
Capacity | 0.125 oz., 4 mL |
Closure Material | Polypropylene |
Closure Size | 13-415 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 16 mm |
Height (English) | 1.61 in. |
Height (Metric) | 41 mm |
Width (English) | 0.62 in. |
Product Type | Diagnostic Bottle |
Capacity (Metric) | 4 mL |
Show More |
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.