Learn More
Macherey-Nagel™ Nano-SIL CN/UV254 HPTLC Plate
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
10 x 10 cm
10 x 20 cm
Unit Size:
Pack of 25
Spécification
Spécification
| Material | Nano-SIL CN |
| Particle Size | 2 to 10 μm |
| Pore Size | 60 Å |
| Quantity | 25 Pk. |
| Type | HPTLC Plate |
| Fluorescent Indicator | UV254 |
| For Use With (Application) | Steroid Hormones, Phenols, Preservatives |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.2 mm |
| Afficher plus |