Learn More
Macherey Nagel Nanocolor Standard Test
Highly sensitive and accurate rectangular cuvettes use large sample volumes for safe and reproducible results.
Brand: Macherey Nagel 91832
Additional Details : Weight : 2.59500kg Transport : UN number : 1888 Chem class : 6.1 Pack group : III
Description
- The use of rectangular cuvettes with 50 mm optical path allows the detection of least amounts of the detectable substance.
- The use of a large sample volume guarantees safe and reproducible results.
- Approved - reliable and comparable results.
- The reaction basis is based on internationally approved standard methods like DIN-, EN-, ISO-, EPA- und APHA.
- Versatile - the perfect test for every request.
- For all important parameters in water and wastewater analysis.
- Precise - easy handling and highest accuracy.
- All tests contain accurate, ready-to-use, predosed reagents.
- Unique - no mix up of tests.
- Every test box is equipped with a color-coded label and can be clearly identified by its color.
Specifications
Anionic detergent | |
40 Determinations | |
Standard Test 1-32. Photometric determination with bromphenol-blue. |
Not seawater suitable | |
10 mm cuvette: 0.1 - 5.0 mg/L MBAS, 50 mm cuvette: 0.02 - 1.00 mg/L MBAS | |
Photometric water analysis test |
Safety and Handling
- NANOCOLOR Detergents anionic
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash with water thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P310-Immediately call a POISON CENTER/doctor/CO2, dry chemical, or foam.
- P330-Rinse mouth.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : chloroform