Learn More
Implen™ NanoPhotometer™ Warranty Expansion
Utilize warranty extensions to protect any NanoPhotometer™ for up to 24 additional months past the date of delivery.
505.00€ - 1010.00€
Specifications
For Use With (Equipment) | NanoPhotometer C40, N50, N60, NP80, N120 |
---|---|
Product Line | NanoPhotometer |
Product Type | Warranty Extension |
Warranty and Service Offering | Extended Warranty |
Product Code | Brand | Duration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Duration | Price | Quantity & Availability | |||||
12825413
|
Implen™
W12 |
12 Months |
505.00€
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
15578319
|
Implen™
W24 |
24 Months |
1010.00€
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
Description
- Warranty extensions protect any NanoPhotometer™
- Choose an extension of either 12 or 24 months
Specifications
NanoPhotometer C40, N50, N60, NP80, N120 | |
NanoPhotometer | |
Warranty Extension | |
Extended Warranty |
For Research Use Only