Learn More
BRAND™ Narrow Neck Glass Erlenmeyer Flasks
These Erlenmeyer Flasks are manufactured from Boro 3.3 glass and are acc. to DIN ISO 1773.
Brand: BRAND™ 92768
69.62 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 1.53000kg
Description
- With beaded rim and graduation
- Narrow neck for controlled pouring and dispensing
- Differing package counts according to capacity
Specifications
DIN ISO 1773 | |
3000 mL | |
310 mm | |
2 | |
Yes |
52 mm | |
Borosilicate Glass (3.3) | |
Narrow | |
Erlenmeyer flask | |
187 mm |