Learn More
Thermo Scientific™ National Microvolume Inserts for 8 x 40mm Shell Vials
Product Code. 12400248
Click to view available options
Volume (Metric):
275 μL
300 μL
Style:
Precision Point Tip
Pulled Point Tip
Unit Size:
Pack of 100
Product Code. 12400248
Brand: Thermo Scientific™ C401596P
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
  • Polyspring™ inserts feature a polyethylene spring to center the bottom of the insert and to provide a cushion against needle contact
  • Precision point inserts permit maximum sample extraction
  • Use metal spring with inserts that lack the Polyspring
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 300 μL
Style Precision Point Tip
Product Line National
For Use With (Equipment) 8 x 40 mm Shell Vials